Learn More
ROBU VitraPOR™ Filter Disc, Untreated, Ground Cylindrical, 210 mm
The faces of the VitraPOR™ standard filter discs are press moulded and may show thickness deviations.
Brand:  ROBU VitraPOR™ 52101
Additional Details : Weight : 1.01650kg
 View more versions of this product
Product Code. 16823146
  • 4646.10 SEK / Each
Estimated Shipment: 10-12-2021
Add to basket
Description
Description
  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Specifications
1 μm
Non-sterile
19.0 ± 4.0 mm
1 Ea.
Borosilicate Glass 3.3
210 mm
1