missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Filter Disc, Untreated, Ground Cylindrical, 300 mm

The faces of the VitraPOR™ standard filter discs are press moulded and may show thickness deviations.

Brand:  ROBU VitraPOR™ 53004

Additional Details : Weight : 2.26100kg

 View more versions of this product

Product Code. 16883186

  • 12580.00 SEK / Each

Estimated Shipment: 17-04-2023
to see stock.
Add to basket

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

Borosilicate Glass 3.3
1 Ea.
26.0 ± 6.0 mm
Non-sterile
4
300 mm
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers