missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Filter Disc, Untreated, Ground Cylindrical, 380 mm

The faces of the VitraPOR™ standard filter discs are press moulded and may show thickness deviations.

Brand:  ROBU VitraPOR™ 53804

Additional Details : Weight : 5.22500kg

 View more versions of this product

Product Code. 16883196

  • 21085.00 SEK / Each

Estimated Shipment: 10-12-2021
Add to basket

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

Borosilicate Glass 3.3
380 mm
4
Non-sterile
38.0 ± 8.0 mm
1 Ea.
Videos
SDS
Documents
Certificates
Special Offers

Special Offers