Learn More
ROBU VitraPOR™ Filter Disc, Untreated, Ground Cylindrical, 40 mm
The faces of the VitraPOR™ standard filter discs are press moulded and may show thickness deviations.
Brand: ROBU VitraPOR™ 15404
Additional Details : Weight : 0.01000kg
Description
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Borosilicate Glass (3.3) | |
10/Pk. | |
4.5 ± 1.0 mm |
Non-sterile | |
4 | |
40 mm |