Learn More
ROBU VitraPOR™ Filter Disc, Untreated, Ground Cylindrical, 40 mm
The faces of the VitraPOR™ standard filter discs are press moulded and may show thickness deviations.
Brand:  ROBU VitraPOR™ 15400
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 12080422
  • 715.00 SEK / Pack of 10
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
Description
Description
  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Specifications
Borosilicate Glass (3.3)
10/Pk.
4.5 ± 1.0 mm
Non-sterile
0
40 mm