Learn More
ROBU VitraPOR™ Filter Disc, Untreated, Ground Cylindrical, 90 mm
The faces of the VitraPOR™ standard filter discs are press moulded and may show thickness deviations.
Brand:  ROBU VitraPOR™ 15904
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12084767
  • 1517.00 SEK / Pack of 5
Estimated Shipment: 20-08-2021
Add to basket
Description
Description
  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Specifications
90 mm
4
Borosilicate Glass 3.3
5/Pk.
6.5 ± 1.5 mm