Learn More
Thermo Scientific™ Barnstead™ Classic Stills Replacement Filter element
Maintain the performance of your Thermo Scientific™ Barnstead™ Classic Still with replacement consumables such as UV lamps and air filters.
Brand: Thermo Scientific™ 25001-DB
Additional Details : Weight : 0.50000kg
Description
Maintain the performance of your still with Thermo Scientific™ Barnstead™ Classic Stills Replacement Consumables, including UV lamp and Thermo Scientific™ Ventgard™ filter element (the consumable portion of the Ventgard filter assembly). Prevent air from entering the reservoir via the condenser atmospheric vent and protect stored water against airborne particulates, organics and CO2 with the Ventgard air filter.
Maintain the performance of your still with Thermo Scientific™ Barnstead™ Classic Stills Replacement Consumables, including UV lamp and Thermo Scientific™ Ventgard™ filter element (the consumable portion of the Ventgard filter assembly). Prevent air from entering the reservoir via the condenser atmospheric vent and protect stored water against airborne particulates, organics and CO2 with the Ventgard air filter.
Specifications
Barnstead Classic Stills | |
Ventguard Filter Element | |
Ventguard Filter Element | |
Fits Ventgard Air Filter |