Learn More
BRAND™ Filter Funnel Supports
Polypropylene plates with aluminum support rod
1945.00 SEK - 2205.00 SEK
Specifications
Material Polypropylene
Quantity 1
View More Specs
Products 2
Product Code Brand Holds Price Quantity & Availability  
Product Code Brand Holds Price Quantity & Availability  
10018020
View Documents Promotion Details
BRAND™
74511
4 funnels
2205.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 15-05-2024
to see stock.
Add to basket
10212091
View Documents Promotion Details
BRAND™
74506
2 funnels
1945.00 SEK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 15-05-2024
to see stock.
Add to basket
Description
Description
  • Manufactured from polypropylene plates with aluminum support rod
  • Adjustable funnel height
  • Support rod is 12.7 mm in diameter, 595 mm long
  • Provided individually
  • Specifications
    Specifications
    Polypropylene
    1