Learn More
Lenz Laborglasinstrumente™ Filter Funnels
Made of borosilicate glass. Lenz Laborglasinstrumente™ Filter funnels are ideal for separating solids from liquids via laboratory filtration.
414.46 SEK - 6587.90 SEK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10284291
|
Lenz Laborglasinstrumente™
07195379 |
4000 mL |
6587.90 SEK
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
10757141
|
Lenz Laborglasinstrumente™
07195328 |
50 mL |
414.46 SEK
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
Description
- Made from borosilicate glass
- Separates solids from liquids
Specifications
Borosilicate Glass | |
3 | |
1 |