Learn More
Lenz Laborglasintrumente Filter Funnels
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 07195334
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10112101
  • 464.28 SEK / Each
Estimated Shipment: 14-06-2024
to see stock.
Add to basket
Description
Description
  • Filter funnels
  • Material: borosilicate glass 3.3
Specifications
Specifications
75 mL
130 mm
3