Learn More
Fisherbrand™ Filter Pump with Non-Return Valve
Fitted with rifled nozzles for water and vacuum connections.
Brand:  Fisherbrand™ 3511
424.92 SEK valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Product Code. 10441904
  • 574.00 SEK / Each
Estimated Shipment: 12-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured in nickel plated brass and stainless steel
  • Extremely robust and durable desgin
  • Fitted with riffled connections, so a rubber tube can be connected to the inlet/outlet
Note: This product, 10441904, is the top item shown in the image above. 
Specifications
Specifications
Non-return valve
Nickel plated brass and stainless steel
Water and vacuum connections
Filter Pump