Learn More
Fisherbrand™ Filter Ring Balance
Designed for use with Welch Ilmvac™ air filters.
Brand:  Fisherbrand™
Additional Details : Weight : 0.01000kg
Product Code. 10511232
  • 331.78 SEK / Each
Estimated Shipment: 26-07-2024
to see stock.
Add to basket
Description
Description
  • Filter Ring Balance
  • 25μm
  • NW25
  • With DN 25 Viton O-ring
Specifications
Specifications
Spare filter ring balance
25 μm