Learn More
Sartorius Stedim Biotech™ Filter Table, Upper Part for 25mm Stainless Steel Filter Holder
Sartorius Stedim Biotech™ Filter Table, Upper Part for 25mm Stainless Steel Filter Holder is suitable as a replacement part for this in-line filtration assembly.
Brand:  Sartorius Stedim Biotech™ 6981031
Additional Details : Weight : 0.09600kg
Product Code. 15972650
  • 2410.00 SEK / Each
In Stock
to see stock.
Add to basket
Description
Description
  • Quality replacement component.
  • Suitable for stainless steel pressure filter holder for a 25mm diameter filter membrane.
Specifications
Specifications
Top part of table
Stainless steel pressure filter holder for 25mm diameter membrane filter. Order No. 16251