Learn More
ROBU VitraPOR™ Filter Thimble, CFE
Filter Thimble with exact glass flange.
Brand:  ROBU VitraPOR™ 20682
Additional Details : Weight : 0.10000kg
Product Code. 10554931
  • 1300.00 SEK / Each
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
Description
Description
  • For many current systems
  • They are highly resistant to most chemicals and temperatures up to 500°C
Specifications
Specifications
1 Ea.
Borosilicate Glass (3.3)
2
34 mm
60 mm