Learn More
Agilent BioTek Filters for ELx800 Absorbance Microplate Reader
Accessory for use with ELx800 Absorbance Microplate Reader.
Brand:  Agilent BioTek 7334450
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10424083
  • 5210.00 SEK / Each
Estimated Shipment: 16-05-2024
to see stock.
Description
Description
Accessory for use with ELx800 Absorbance Microplate Reader.
Specifications
Specifications
Filter
ELx800 Absorbance Microplate Readers
450 nm