Läs mer
Finafloxacin, MedChemExpress
Produktkod. 16405698
Encompass_Preferred
Change view
Kvantitet:
1 mg
5 mg
10 mg
50 mg
Förpackningsstorlek:
1mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16405698 10 mg -
16471349 1 mg 1mg
16431857 50 mg -
16448387 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16405698 Leverantör Medchem Express Leverantörsnummer HY13451
MedChemExpress Finafloxacin is a fluoroquinolone antimicrobial agent that exhibits optimum efficacy in slightly acidic environments.
Finafloxacin is a fluoroquinolone antimicrobial agent that exhibits optimum efficacy in slightly acidic environments.
Specifikationer
Kemiskt namn eller material Finafloxacin
CAS 209342-40-5
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C20H19FN4O4
Kvantitet 10 mg
Löslighetsinformation DMSO : 6.4 mg/mL (16.06 mM; Need ultrasonic and warming)
LEDER O=C(C1=CN(C2CC2)C3=C(C=C(F)C(N4C[C@]5([H])OCCN[C@@]5([H])C4)=C3C#N)C1=O)O
Molekylvikt (g/mol) 398.39
Formel vikt 398.39
Procent renhet 99.87%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation