Läs mer
Finasteride 2-(2-Methylpropanol)amide, TRC
Produktkod. 30136696
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136696 10 mg
30135920 50 mg
30012563 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136696 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF34202010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 116285-36-0
Molekylformel C23 H36 N2 O3
Molekylvikt (g/mol) 388.54
Synonym 1H-Indeno[5,4-f]quinoline-7-carboxamide, 2,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-N-(2-hydroxy-1,1-dimethylethyl)-4a,6a-dimethyl-2-oxo-, (4aR,4bS,6aS,7S,9aS,9bS,11aR)-, 4-Azaandrost-1-ene-17-carboxamide, N-(2-hydroxy-1,1-dimethylethyl)-3-oxo-, (5α,17β)-, (4aR,4bS,6aS,7S,9aS,9bS,11aR)-2,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-Tetradecahydro-N-(2-hydroxy-1,1-dimethylethyl)-4a,6a-dimethyl-2-oxo-1H-indeno[5,4-f]quinoline-7-carboxamide, 1H-Indeno[5,4-f]quinoline-7-carboxamide, 1,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-N-(2-hydroxy-1,1-dimethylethyl)-4a,6a-dimethyl-2-oxo-, [4aR-(4aα,4bβ,6aα,7α,9aβ,9bα,11aβ)]-, Finasteride 2-(2-Methylpropanol)amide, (5α,17β)-N-(1-Hydroxymethyl-1-methylethyl)-3-oxo-4-aza-5-androst-1-ene-17-carboxamide
IUPAC-namn (1S,3aS,3bS,5aR,9aR,9bS,11aS)-N-(1-hydroxy-2-methylpropan-2-yl)-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxamide
LEDER CC(C)(CO)NC(=O)[C@H]1CC[C@H]2[C@@H]3CC[C@H]4NC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material Finasteride 2-(2-Methylpropanol)amide
Kvantitet 10 mg
InChI formel InChI=1S/C23H36N2O3/c1-21(2,13-26)25-20(28)17-7-6-15-14-5-8-18-23(4,12-10-19(27)24-18)16(14)9-11-22(15,17)3/h10,12,14-18,26H,5-9,11,13H2,1-4H3,(H,24,27)(H,25,28)/t14-,15-,16-,17+,18+,22-,23+/m0/s1
Formel vikt 388.2726