Läs mer
Tocris Bioscience™ Finasteride
Beskrivning
Antiandrogen that inhibits type II 5α reductase (IC50 = 65 nM). Suppresses the conversion of testosterone to dihydrotestosterone. Reduces prostatic dihydrotestosterone levels and prostate size in vivo. Orally active.
Specifikationer
Specifikationer
| Kemiskt namn eller material | Finasteride |
| CAS | 98319-26-7 |
| Kvantitet | 50 mg |
| Molekylformel | C23H36N2O2 |
| Formel vikt | 372.54 |
| Renhet | >99% |
| Molekylvikt (g/mol) | 372.553 |