missing translation for 'onlineSavingsMsg'
Läs mer

Fingolimod Stearoate Amide, TRC

Produktkod. 30137765
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137765 1 g
30004881 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30137765 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF3420551G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 1242271-27-7
Molekylformel C37H67NO3
Molekylvikt (g/mol) 573.93
Synonym N-(1-Hydroxy-2-(hydroxymethyl)-4-(4-octylphenyl)butan-2-yl)stearamide
LEDER Cl.C[C@@H](N)c1c(F)cccn1

Specifikationer

Kemiskt namn eller material Fingolimod Stearoate Amide
Kvantitet 1 g
InChI formel InChI=1S/C37H67NO3/c1-3-5-7-9-11-12-13-14-15-16-17-18-19-21-23-25-36(41)38-37(32-39,33-40)31-30-35-28-26-34(27-29-35)24-22-20-10-8-6-4-2/h26-29,39-40H,3-25,30-33H2,1-2H3,(H,38,41)
Formel vikt 573.5121
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.