missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FIP1L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | FIP1L1 |
---|---|
Concentration | 0.24 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16879893
|
Proteintech
12276-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16869893
|
Proteintech
12276-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
FIP1L1 is a component of the cleavage and polyadenylation specificity factor (CPSF) complex that plays a key role in pre-mRNA 3′-end formation, recognizing the AAUAAA signal sequence and interacting with poly(A) polymerase and other factors to bring about cleavage and poly(A) addition. FIP1L1 contributes to poly(A) site recognition and stimulates poly(A) addition. It binds to U-rich RNA sequence elements surrounding the poly(A) site. FIP1L1 may act to tether poly(A) polymerase to the CPSF complex.Specifications
FIP1L1 | |
Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
Q5U317, Q6UN15, Q9D824 | |
289582, 66899, 81608 | |
FIP1L1 Fusion Protein Ag2921 | |
Primary | |
-20°C | |
FIP1L1 |
0.24 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Factor interacting with PAP, FIP1, FIP1 like 1 (S. cerevisiae), FIP1 like 1 protein, FIP1L1, FIP1L1 PDGFRalpha, hFip1, Rhe | |
Fip1l1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |