missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FIR Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565956
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: VEESDNEWSVP HCFTIYAAQK TIVVAASTRL EKEKWMLDLN SAIQAAKSGG DTAPALPGRT VCTRPPRSPN EVSLEQESED DARGVR Highest antigen sequence identity to the following orthologs - mouse 75%, rat 75%.
FIR Polyclonal antibody specifically detects FIR in Human samples. It is validated for Immunocytochemistry.Specifications
FIR | |
Polyclonal | |
Unconjugated | |
FARP2 | |
AI465173; BC009153; D030026M03Rik; Farp2; FERM domain including RhoGEF; FERM domain-including RhoGEF; FERM, ARH/RhoGEF and pleckstrin domain protein 2; FERM, ARHGEF and pleckstrin domain-containing protein 2; FERM, RhoGEF and pleckstrin domain protein 2; FERM, RhoGEF and pleckstrin domain-containing protein 2; FGD1-related Cdc42-GEF; Fir; FRG; Kiaa0793; mKIAA0793; PH domain-containing family C member 3; pleckstrin homology domain-containing family C member 3; PLEKHC3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9855 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O94887 | |
FARP2 | |
Recombinant Human FIR | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only