Läs mer
Fisher BioReagents™ SOC Medium
:
5 x 10mL
Beskrivning
- Nutrient-rich growth media used to maximize the transformation efficiency of competent E. coli cells
- Liquid media is conveniently packaged and supplied as 5 x 10mL bottles
Specifikationer
Specifikationer
| Form | Liquid |
| Tillverkningsursprung | United States |
| Medietyp | SOC Medium |
| Fyll volym | 10 mL |
| Formulering | Casein Peptone 20g/L, Yeast extract, 5g/L, Sodium Chloride 0.58g/L; Potassium Chloride 0.19g/L, Magnesium Chloride 0.95g/L, Magnesium Sulfate 1.20g/L, Glucose 3.60g/L |
| Förpackningstyp | Bottle |
| Kvantitet | 5 x 10 mL |