Learn More
Fisherbrand™ Elite™ Multichannel Pipetters
New, improved ergonomic design provides newly designed finger rest and index finger and hand support for maximum comfort
5169.00 SEK - 5927.00 SEK
Product Code | Brand | Volume (Metric) | Number of Channels | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Number of Channels | Price | Quantity & Availability | ||||
11835772
|
Fisherbrand™
|
5 to 50 μL | 8 |
5274.00 SEK
Each |
06-04-2021 |
11845772
|
Fisherbrand™
|
30 to 300 μL | 8 |
5169.00 SEK
Each |
|
11815772
|
Fisherbrand™
|
1 to 10 μL | 8 |
5274.00 SEK
Each |
17-02-2021 |
11885772
|
Fisherbrand™
|
30 to 300 μL | 12 |
5927.00 SEK
Each |
|
Description
- Fully autoclavable - entire pipetter can be steam autoclaved
- Soft-touch tip ejection—a gearing mechanism reducing tip ejection force up to 50%
- Ergonomic comfortable lightweight handle - comfortable grip and effortless tip attachment
- Optimal fit with SureOne pipetter tips - best pipetting performance
- Super blow-out piston - creates 150% bigger air push than conventional piston for delivery of the micro size drops
- 8- and 12-channel pipetters with volume range of 1 up to 300μL
- Extremely low plunger forces - work longer without fatigue and reduce the risk of RSI