Learn More
Fisherbrand™ Filter/Regulator and Lubricator Complete Assembly for Air Stirrers
For long and trouble-free life of air-powered stirrers.
Brand:  Fisherbrand™
Additional Details : Weight : 1.00000kg
Product Code. 15182997
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Filter removes oil and water aerosols and foreign particles larger than 40 microns in the air line
  • Integral filter/regulator and lubricator combination
  • With 1.5 in. diameter gauge, 0-160 psi
Order Info
  • Automatic air line lubricator must be installed in the air line just ahead of the motor
  • Lubricator should be adjusted to feed one drop of oil for every 50-70 cfm of air groung through the motor (approximately 1 drop every five minutes)
Specifications
Specifications
Air Stirrer
Long and trouble-free life of air-powered stirrer
1.5 in. dia. Gauge
Lubricator/Filter/Regulator Assembly