Learn More
Fisherbrand™ Filter/Regulator and Lubricator Complete Assembly for Air Stirrers
For long and trouble-free life of air-powered stirrers.
Brand: Fisherbrand™
Additional Details : Weight : 1.00000kg
Description
Filter removes oil and water aerosols and foreign particles larger than 40 microns in the air line
- Integral filter/regulator and lubricator combination
- With 1.5 in. diameter gauge, 0-160 psi
Order Info
- Automatic air line lubricator must be installed in the air line just ahead of the motor
- Lubricator should be adjusted to feed one drop of oil for every 50-70 cfm of air groung through the motor (approximately 1 drop every five minutes)
Specifications
Air Stirrer | |
Long and trouble-free life of air-powered stirrer | |
1.5 in. dia. Gauge | |
Lubricator/Filter/Regulator Assembly |