Läs mer
Fisherbrand™ Isotemp™ Hot Plate, Ambient to 300°C, Aluminum
Produktkod. 15373518 Handla allt Fisherbrand Traceable Produkter
Change view
Mått (L x B x H):
13 x 8,2 x 3,8 tum (33 x 20,8 x 9,7 cm)
Förpackningsstorlek:
Varje
Produktkod. Mått (L x B x H) Förpackningsstorlek
15373518 13 x 8,2 x 3,8 tum (33 x 20,8 x 9,7 cm) Varje
1 options
Produktkod. 15373518
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Shape: Square; Size: 7.25 x 7.25 in.; Plug Type: EU-Style
Experience exceptional safety and temperature performance for your routine protocols. Available in three sizes, providing flexibility from microscale chemistries to production operations.
Advanced technology
  • Strong magnetic coupling
  • Temperature adjustable in 1 degree increments
  • HOT TOP warning system designed to protect from accidental burns with prominent display when heating surface is above 50°C (122°F)
  • Automatic over-temperature protection
Flexible, robust design
  • Top plate choices in ceramic or aluminum
  • Raised display design protects electronics from spills
  • Optional temperature probe and splash guard protection shield
  • Variable sizes for different sample volumes
Certifikat
cCSAus (100-120V models), CE (230V models)
Uppfyllande
RoHS compliant
Garanti
Three years
TRUSTED_SUSTAINABILITY
Specifikationer
Beskrivning Hotplate
Material för toppplatta Aluminum
Storlek 7.25 x 7.25 in.
Temperatur (metrisk) Ambient to 300°C
Höjd (engelska) 3.8 in.
Längd (engelska) 13 in.
Bredd (engelska) 8.2 in.
Höjd (metrisk) 9.7 cm
Längd (metrisk) 33 cm
Bredd (metrisk) 20.8 cm
Elektriska krav 230 V, 50/60 Hz
Frekvens 50/60 Hz
Pluggtyp EU
Form Square
Spänning 230 V
Typ Hot Plate
Certifikat och efterlevnad CE, ROHS Compliant
Garanti 3 Years
Mått (L x B x H) 13 x 8,2 x 3,8 tum (33 x 20,8 x 9,7 cm)