Learn More
Fisherbrand™ Jack Stand
Product Code. 12951161 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
Model 505/705 Sonic Dismembrator
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
12951161 Model 505/705 Sonic Dismembrator Each
1 options
Product Code. 12951161
  • ONLINE EXCLUSIVE PRICE
    2935.00 SEK
    2313.80 SEK / Paket med 1
    Save 621.20 SEK
    21% Av
Beräknad leverans: 16-12-2026
Add to basket
Add to basket
Ideal for raising samples up to a stationary probe mounted within the enclosure or on a stand.
Fisher Scientific™ Jack Stand is ideal for samples to a stationary probe mounted within the enclosure.
  • Platform that raises and lowers samples under a stationary probe
  • For use with Model 505/705 Sonic Dismembrator.
Specifications
Product Type Jack Stand
Dimensions (L x W) 6 x 6 in. platform
For Use With (Application) Ideal to raise samples to a stationary probe mounted in an enclosure
For Use With (Equipment) Model 505/705 Sonic Dismembrator
Packaging Quantity 1 Ea.