Learn More
Fisherbrand™ Wellwash™ and Wellwash™ Versa™ Microplate Strip Washer: Covers
Accessories for Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
Brand: Fisherbrand™
Additional Details : Weight : 0.01000kg
Description
For use with Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
Aerosol cover prevents aerosols of infectious diseases from spreading
ELISA-based assays, cell washing
Specifications
ELISA-based assays, cell washing | |
Aerosol Cover | |
Each | |
Wellwash Microplate Strip Washer |