Learn More
Fisherbrand™ 20-Tooth Combs for Horizontal Midigel Systems
20-Tooth Combs for Horizontal Midigel Systems
Brand: Fisherbrand™
Additional Details : Weight : 0.80000kg
Description
- Accessory for use with Horizontal Midigel Systems
Specifications
Owl™ A1 Large Gel Electrophoresis System | |
9 x 11 cm | |
0.15 cm | |
0.17 in. |
Standard Comb | |
20 | |
30 μL | |
0.45 cm |