Learn More
Fisherbrand™ Horizontal Large Gel Systems: Marker
Preparative/marker for 1.5mm comb
Brand: Fisherbrand™
Additional Details : Weight : 0.80000kg
Specifications
Owl™ A2 and A5 Large Gel Electrophoresis System | |
2 | |
1450/35 μL | |
19.4/0.5 cm |
Preparative Comb | |
0.15 cm | |
7.6/0.19 in. |