Learn More
Fisherbrand™ Adhesive Entryway Mats
Trap contaminating dirt, dust and bacteria before they invade cleanrooms
402.95 SEK - 638.60 SEK
Products 2
Product Code Brand Surface Coating Includes Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Surface Coating Includes Dimensions (L x W) Price Quantity & Availability  
11803191 View Documents Fisherbrand™
Low-tack 40 clear sheets on white mat 24 x 45 in. 638.60 SEK
Pack of 120 View alternative products
Save Up to 
»
Estimated Shipment
27-02-2020
Add to basket
12795955 View Documents Fisherbrand™
Low-tack 40 clear sheets on white mat 24 x 45 in. 402.95 SEK
Pack of 4 View alternative products
Save Up to 
»
Call for latest availability.
+46 (0)31 352 32 30
Add to basket
Description
Description
  • Clear 2mil thick polyethylene sheets coated with environmentally friendly waterborne acrylic adhesive
  • Pull-tab system makes it easy to peel off top sheet when it becomes soiled; remaining sheets stay intact and clean
  • Will not curl, tear or wrinkle
  • Adhesive will not transfer to footwear
  • For standard tack needs: standard tack, 30; ideal for use in Electronic, Pharmaceutical, Laboratory and Food industry cleanrooms
Order Info
Sheets per mat: Low tack, 40; High tack, 30.