Learn More
Fisherbrand™ Advanced Protection Disposable Facemasks
Provides convenient protection
Brand:  Fisherbrand™
Additional Details : Weight : 2.21840kg
 View more versions of this product
Product Code. 15481932
  • 332.00 SEK / Pack of 500
Estimated Shipment
03-08-2018
Add to basket
Description
Description
  • Advanced protection
  • Disposable, single use
  • Choice of ear loops or tie closure
  • Nonwoven polypropylene outer layer
  • Thermally bonded nonwoven polypropylene inner layer
  • Latex-free
  • Choice of blue or white
  • Universal size/fit
Specifications
Specifications
Blue
Universal
Yes
Nonwoven Polypropylene (outer and inner) / Meltblown Polypropylene (filter)
PP (Polypropylene)þPP (Polypropylene)
Yes
3-ply
Tie closures
Videos
Documentation