Learn More
Fisherbrand™ Advanced Protection Disposable Facemasks
Provides convenient protection
Brand:  Fisherbrand
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15451932
  • 261.00 SEK / Pack of 500
Estimated Shipment
06-03-2018
Add to basket
Description
Description
  • Advanced protection
  • Disposable, single use
  • Choice of ear loops or tie closure
  • Nonwoven polypropylene outer layer
  • Thermally bonded nonwoven polypropylene inner layer
  • Latex-free
  • Choice of blue or white
  • Universal size/fit
Specifications
Specifications
White
Universal
Yes
Nonwoven Polypropylene (outer) / Thermally Bonded Nonwoven Polypropylene (inner)
PP (Polypropylene)
Yes
2-ply
Earloops
Videos
Documentation