Learn More
Fisherbrand™ Basic Protection Polypropylene Disposable Lab Coats with Elastic Cuffs
Latex-free spunbond polypropylene construction lab ware
Brand:  Fisherbrand™
214.50 SEK valid until 2018-08-31
Use promo code "13204" to get your promotional price.
Additional Details : Weight : 1.20000kg
 View more versions of this product
Product Code. 15471922
  • 275.00 SEK / Pack of 25
Sweden Stock
(immediate availability): 2
Add to basket
Description
Description
  • Single-use lab coat
  • Standard collar, no pockets
  • Packed 25 lab coats/case
  • Elastic cuffs hold sleeves in place
  • Five snaps for a secure closure
  • Fabric weight: 50g/m2
  • White
Note: Other materials, colors and options are available. See Fisherbrand Basic Protection Disposable Lab Coats with Pockets, and Fisherbrand Basic Protection Disposable SMS Lab Coats in a variety of colors.
Specifications
Specifications
Standard
White
Polypropylene
2X-Large
Yes
Snap
Elastic
0
5-Snap
Videos
Documentation Special Offers
Special Offers
  • Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    : 13204
    2018-08-31
    *Discount will apply to qualifying orders received by Fisher Scientific between 04/06/2018 and 31/08/2018. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions