Learn More
Fisherbrand™ Combs for Mini-Plus Wide Format Horizontal Gel Units, 2mm Thick
For use with Mini-Plus Wide Format Horizontal Gel Units.
Brand: Fisherbrand™
Additional Details : Weight : 0.01000kg
Description
- Recommended to scale-up nucleic acid volumes for preparatory techniques
- To maximise sample volume
- Height-adjustable without any requirement for specialist tools or comb holders, to give user full control over well depth and sample-loading volume
- Rigid comb back prevents heat-induced warping
- Reversible loading guides sit directly above each well to provide a convenient loading template for single- and multi-channel pipets
- 8-sample and 16-sample combs are Multichannel pipettor compatible
Specifications
Blue | |
Comb | |
1 Each | |
58 μL |
Mini-Plus Wide Format Horizontal Gel Units | |
16 | |
2 mm |