Learn More
Fisherbrand™ Combs for SUB-GEL Mini Horizontal Gel Electrophoresis Units, 0.75mm Thick
For use with SUB-GEL Mini Horizontal Gel Electrophoresis Units.
Brand:  Fisherbrand™
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11817563
  • 201.00 SEK / Each
Estimated Shipment: 01-07-2024
to see stock.
Add to basket
Description
Description
  • Recommended for high resolution gels and publication quality data
  • For tightly resolved bands
  • Height-adjustable without any requirement for specialist tools or comb holders, to give user full control over well depth and sample-loading volume
  • Rigid comb back prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single- and multi-channel pipets
  • 8-sample and 12-sample combs are Multichannel pipettor compatible
Specifications
Specifications
Black
Comb
1 Each
19 μL
Sub-Gel Mini Horizontal Gel Electrophoresis Unit
8
0.75 mm