Läs mer
Fisherbrand™ Finnpipette™ II Digital Multichannel Pipetters
Produktkod. 11827511 Handla allt Fisherbrand Produkter
Change view
Volym (metrisk):
5 till 50 μl
50 till 300 μl
Antal kanaler:
8
12
Färg:
Gult
Orange
Ökar:
0,5 μl
5 μl
Förpackningsstorlek:
Varje
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Volym (metrisk) Antal kanaler Färg Ökar unitSize
11827511 50 till 300 μl 8 Gult 5 μl Varje
11553312 5 till 50 μl 8 Orange 0,5 μl Varje
11593312 50 till 300 μl 12 Gult 5 μl Varje
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 11827511
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Advanced pipetting performance in a lightweight, ergonomic design
  • 8- and 12-channel pipetters, ideal for multiplate applications
  • Minimum force allows tips to be ejected with just a gentle push
  • Reverse pipetting mode allows pipetting of viscous and surface-active solutions
  • Made from inert, biologically inactive polymers
  • Easy to disassemble for routine maintenance and cleaning
  • Serial number and calibration certificate included
  • Easy, in-lab calibration
Superior Design
  • Rounded hand grip and modified grippy finger rest ensure secure grip and correct pipetting angle; help minimize hand fatigue
  • Large digital display is easy to read, even when pipetting
  • Large color-coded knob makes it easy to set and identify desired volume; fast-dial system allows 0.1μL fine adjustment
  • Separate soft-touch tip ejector relieves hand stress, prevents accidental detipping
  • Super blowout feature on micro models ensures highest level of accuracy and precision at low-volume settings
Beställningsinfo
For optimum performance, use Fisherbrand SureOne™ Pipet Tips or other universal fit pipet tips.
Garanti
Three years
TRUSTED_SUSTAINABILITY
Specifikationer
Autoklaveringsbar Autoclavable
Färg Gult
Antal kanaler 8
Produktlinje Finnpipette™ II
Typ Digital Pipetter
Volym (metrisk) 50 till 300 μl
Ökar 5 μl
Garanti 3 Years