Learn More
Fisherbrand™ Nonsterile Cotton Balls
Use these cotton balls for a wide range of applications. Fisherbrand™ Nonsterile cottonballs are available in a variety of sizes.
Brand: Fisherbrand™
Additional Details : Weight : 2.62800kg
Description
Use these cotton balls for a wide range of applications. Fisherbrand™ Nonsterile cottonballs are available in a variety of sizes.
- Uniform in size and shape
- Made of pure, natural, bleached cotton for increased absorbency
- Available in sizes medium, large, extra large and bulk roll
Specifications
Cotton Balls | |
Large | |
3.75 lb. |
Non-sterile | |
2000 Cs. | |
0.004 kg |