Läs mer
FISHERBRAND™ FISHERBRAND™ Round Cuvets
Produktkod. 16333416 Handla allt Fisherbrand Traceable Produkter
Change view
Diameter (metrisk):
19 mm
Höjd (metrisk):
105 mm
Produktkod. Diameter (metrisk) Höjd (metrisk)
16333416 19 mm 105 mm
1 options
Produktkod. 16333416
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
FISHERBRAND™ Round Cuvets are designed for use with commercial colorimeters and spectrophotometers. Manufactured from borosilicate glass with a reference mark near the top for accurate repositioning, these cuvets are available in multiple sizes and sold in cases of 36.
For use with commercial colorimeters and spectrophotometers.
  • Reference mark near top assists accurate repositioning of the cuvet
  • Manufactured from borosilicate glass for chemical resistance and optical clarity
  • Compatible with commercial colorimeters and spectrophotometers
  • Available in multiple diameter and height configurations to suit different instruments
  • Sold in cases of 36
Specifikationer
Typ Macro Spectrophotometer Cuvette
Material Borosilicate Glass
Höjd (metrisk) 105 mm
För användning med (utrustning) Coleman Jr. II
För användning med (applikation) Spectrophotometer Calibration
Form Round
Diameter (metrisk) 19 mm