Learn More
Fisherbrand™ Stainless Steel Bow Tongs
Designed using high quality components these corrosion resistant, non-magnetic bow tongs feature excellent heat resistance and great cryogenic stability.
137.00 SEK - 156.00 SEK
Products 2
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
11543522
View Documents Promotion Details
Fisherbrand™
150 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11557872
View Documents Promotion Details
Fisherbrand™
203 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • 304 spec 18/8 non-magnetic stainless steel laboratory bow tongs with PTFE coated jaws
  • Corrosion resistant, non-magnetic bow tongs
  • Applied as a self priming single coat PTFE
  • Excellent heat resistance and great cryogenic stability
  • Excellent non-wetting properties that makes cleaning very easy and can also undergo steam or chemical sterilization
  • The coefficient of friction is generally in the range of 0.05 to 0.20