Learn More
Fisherbrand™ Stainless Steel Bow Tongs
Designed using high quality components these corrosion resistant, non-magnetic bow tongs feature excellent heat resistance and great cryogenic stability.
137.00 SEK - 156.00 SEK
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
11543522
|
Fisherbrand™
|
150 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11557872
|
Fisherbrand™
|
203 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- 304 spec 18/8 non-magnetic stainless steel laboratory bow tongs with PTFE coated jaws
- Corrosion resistant, non-magnetic bow tongs
- Applied as a self priming single coat PTFE
- Excellent heat resistance and great cryogenic stability
- Excellent non-wetting properties that makes cleaning very easy and can also undergo steam or chemical sterilization
- The coefficient of friction is generally in the range of 0.05 to 0.20