Learn More
Fisherbrand™ Mini-Centrifuge 220-230V, 50/60Hz Universal Plug, Grey
Tool-free rotors and quick-spin options make the Fisherbrand Mini-Centrifuge fast and easy to use. The locking lid is functionally designed to fit in the palm of the hand for easy repeat opening and quick tube extraction. 
Brand:  Fisherbrand™
1334.91 SEK valid until 2019-02-28
Use promo code "13474" to get your promotional price.
Additional Details : Weight : 1.50000kg
Product Code. 15358266
  • 2348.95 SEK / Each
Sweden Stock
(immediate availability): 19
Remote Stock
(10 days availability): 63
Add to basket
Description
Description
  • A locking lid for added protection when the unit is in use
  • A larger lid to accommodate longer and skirted tubes, as well as taller tube lids
  • A braking function to quickly stop the unit when powered off or when the lid is opened
  • Meets the latest applicable North American & EU safety requirements
  • Weighted rubberized base eliminates potential movement and further dampens vibration and noise
Specifications
Specifications
Rotor 1: 6 x 1.5/2mL tubes (adapters for 0.5mL and 0.2ml tubes)
Rotor 2: 2 places for 8 x 0.2mL strips or 16 single tubes
2,000 x g
50-60
128mm
220-230V 50/60Hz
6000rpm
No
220-230V
Two rotors, tube rack, storage case, O-rings, tube adapters and low voltage, double insulated power adapter with four interchangeable plugs
Yes
Videos
Documentation View accessory products