Learn More
ACROBiosystems FITC-Labeled Human B7-H3 (4Ig) / B7-H3b Protein, His Tag (MALS verified)
Brand:  ACROBiosystems B7B-HF2E7-25ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 17216060
  • 4272.53 SEK / 25µg
Estimated Shipment: 18-06-2024
to see stock.
Description
Description
Human B7 homolog 3 (B7-H3) is a member of the B7 family of immune proteins that provide signals for the regulation of immune responses. Other family members include B7-1, B7-2, B7-H1/PD-L1, B7-H2, and PD-L2. B7 family proteins are type I transmembrane immunoglobulin (Ig) superfamily members that contain extracellular Ig V‐like and Ig C‐like domains with a short cytoplasmic tail. Termed 4IgB7-H3 or B7-H3b, this molecule has two additional Ig-like domains (one V‐type and one C‐type) and shows a ubiquituous expression pattern.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 48.5 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifications
Specifications
NP_001019907.1
25 μg
Gly 27 - Thr 461
4 Ig-B7-H3,B7-H3,CD276,PSEC0249,UNQ309/PRO352,B7 homolog 3
FITC
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE.
48.5 kDa
FITC-Labeled Human B7-H3 (4 Ig), His Tag (B7B-HF2E7) is expressed from human 293 cells (HEK293). It contains AA Gly 27 - Thr 461 (Accession # Q5ZPR3-1). It is the FITC labeled form of Human B7-H3 (4 Ig), His Tag (Cat. No.B7B-H52E7).
-20°C, aviod light
B7-H3 (4 Ig)
Human
PBS, pH7.4
HEK293
Fluorescent Labeled Protein