Learn More
ACROBiosystems FITC-Labeled Human CD47 Protein, His Tag
4272.53 SEK - 29373.66 SEK
Specifications
Accession Number | NP_942088 |
---|---|
Molecular Weight (g/mol) | 15.6 kDa |
Source | FITC-Labeled Human CD47, His Tag (CD7-HF2H3) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 139 (Accession # NP_942088). |
Immunogen | Gln 19 - Pro 139 |
Storage Requirements | -20°C, avoid light |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17226120
|
ACROBiosystems
CD7-HF2H3-200UG |
200 μg |
29373.66 SEK
200µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17236120
|
ACROBiosystems
CD7-HF2H3-25UG |
25 μg |
4272.53 SEK
25µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Leukocyte surface antigen CD47 is also known as Antigenic surface determinant protein OA3, Integrin-associated protein (IAP) and Protein MER6. CD47 contains 1 Ig-like V-type (immunoglobulin-like) domain. CD47 is very broadly distributed on normal adult tissues. CD47 has a role in both cell adhesion by acting as an adhesion receptor for THBS1 on platelets, and in the modulation of integrins and plays an important role in memory formation and synaptic plasticity in the hippocampus by similarity. CD47 is the receptor for SIRPA, binding to which prevents maturation of immature dendritic cells and inhibits cytokine production by mature dendritic cells. CD47 Interaction with SIRPG mediates cell-cell adhesion, enhances superantigen-dependent T-cell-mediated proliferation and costimulates T-cell activation.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.6 kDa.Specifications
NP_942088 | |
FITC-Labeled Human CD47, His Tag (CD7-HF2H3) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Pro 139 (Accession # NP_942088). | |
-20°C, avoid light | |
CD47 | |
Human | |
HEK293 | |
Fluorescent Labeled Protein |
15.6 kDa | |
Gln 19 - Pro 139 | |
CD47,MER6,IAP,OA3 | |
FITC | |
His Tag | |
Powder |