Läs mer
Tocris Bioscience™ FK 866
Beskrivning
FK 866 is a non-competitive and high-affinity inhibitor of NAMPT (nicotinamide phosphoribosyltransferase, PBEF1) (Ki = 0.3 nM); inhibits NAD biosynthesis. Induces delayed cell death by apoptosis in HepG2 human liver carcinoma cells (IC50 ~1 nM). Induces apoptosis in four different neuroblastoma cell lines; also induces autophagy in SH-SY5Y cells. Potentiates the cytotoxic effects induced by Etoposide (Cat. No. 1226) and Cisplatin (Cat. No. 2251).
Specifikationer
Specifikationer
| Kemiskt namn eller material | (2E)-N-[4-(1-Benzoyl-4-piperidinyl)butyl]-3-(3-pyridinyl)-2-propenamide |
| CAS | 658084-64-1 |
| Kvantitet | 10 mg |
| Mål | NAMPT Inhibitors |
| Molekylformel | C24H29N3O2 |
| Renhet | 98% |