missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11755-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
FK506- and rapamycin-binding proteins (FKBPs) constitute a family of receptors for the two immunosuppressants which inhibit T-cell proliferation by arresting two distinct cytoplasmic signal transmission pathways. PPIases accelerate the folding of proteins.Specifications
FKBP3 | |
Polyclonal | |
Unconjugated | |
FKBP3 | |
25 kDa FK506 binding protein, 25 kDa FKBP, FK506 binding protein 3, FK506 binding protein 3, 25kDa, FKBP 25, FKBP 3, FKBP25, FKBP3, Immunophilin FKBP25, PPIase, PPIase FKBP3, Rotamase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2287, 299104, 30795 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q00688, Q62446 | |
Fkbp3 | |
FKBP3 Fusion Protein Ag2351 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |