missing translation for 'onlineSavingsMsg'
Läs mer

Abnova™ FKBP3 Recombinant Protein

Produktkod. 16105021
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 μg
Förpackningsstorlek:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16105021 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16105021 Leverantör Abnova™ Leverantörsnummer H00002287P01.10ug

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Human FKBP3 full-length ORF with GST-tag at N-terminal

The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. This encoded protein is a cis-trans prolyl isomerase that binds the immunosuppressants FK506 and rapamycin, as well as histone deacetylases, the transcription factor YY1, casein kinase II, and nucleolin. It has a higher affinity for rapamycin than for FK506 and thus may be an important target molecule for immunosuppression by rapamycin.

  • Theoretical MW: 50.38kDa
  • Preparation Method: In vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 Fast Flow
  • Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifikationer

För användning med (applikation) Antibody Production, ELISA, Protein Array, Western Blot
Formulering 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Molekylvikt (g/mol) 50.38
Namn Human FKBP3 Full-length ORF Recombinant Protein with GST-tag at N-terminal
pH-intervall 8
Beredningsmetod In vitro wheat germ expression system
Reningsmetod Glutathione Sepharose 4 Fast Flow
Kvalitetskontrolltestning 12.5% SDS-PAGE stained with Coomassie Blue
Kvantitet 10 μg
Källa Wheat Germ (in vitro)
Förvaringskrav -80°C
Korsreaktivitet Human
Rekombinant Recombinant
Form Solution
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.