missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP8 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11173-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
FKBP8 is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. Unlike the other members of the family, it does not seem to have PPIase/rotamase activity. It may have a role in neurons associated with memory function.The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. Unlike the other members of the family, this encoded protein does not seem to have PPIase/rotamase activity. It may have a role in neurons associated with memory function.Specifications
FKBP8 | |
Polyclonal | |
Unconjugated | |
FKBP8 | |
38 kDa FK506 binding protein, 38 kDa FKBP, FK506 binding protein 8, FK506 binding protein 8, 38kDa, FKBP 38, FKBP 8, FKBP38, FKBP8, FKBPr38, hFKBP38, PPIase FKBP8, Rotamase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14232, 23770 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35465, Q14318 | |
Fkbp8 | |
FKBP8 Fusion Protein Ag1649 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |