missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18073-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
FKBP9 is a PPIase which accelerate the folding of proteins during protein synthesis.Specifications
FKBP9 | |
Polyclonal | |
Unconjugated | |
FKBP9 | |
63 kDa FK506 binding protein, 63 kDa FKBP, FK506 binding protein 9, FKBP 63, FKBP 9, FKBP60, FKBP63, FKBP9, PPIase, PPIase FKBP9, Rotamase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11328, 27055, 297123 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95302, Q66H94, Q9Z247 | |
FKBP9 | |
FKBP9 Fusion Protein Ag12715 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |