Learn More
Zeiss™ FL Attenuator
Insert this discrete attenuator into the aperture stop plane of a reflected-light illuminator. Filter wheel with 6 positions: Pos. 1: clear aperture; Pos. 2-6: transmission appr. 70, 50, 40, 20 and 2%
Brand: Zeiss™ 423730-9130-000
Additional Details : Weight : 0.01000kg
Description
Compatible Microscopes:
- Axio Examiner.D1
- Axio Examiner.Z1
- Axio Scope.A1
- Axio Vert.A1 FL
- Axio Vert.A1 MAT
Specifications
Slider FL Attenuator | |
Filter wheel with 6 positions | |
14 x 40 mm |
For insert into the aperture stop plane of reflected-light illuminator | |
1 Ea. |