Learn More
Zeiss™ FL Attenuator
Insert this discrete attenuator into the aperture stop plane of a reflected-light illuminator. Filter wheel with 6 positions: Pos. 1: clear aperture; Pos. 2-6: transmission appr. 70, 50, 40, 20 and 2%
Brand:  Zeiss™ 423730-9130-000
Additional Details : Weight : 0.01000kg
Product Code. 11815450
  • 4367.41 SEK / Each
Estimated Shipment: 20-05-2024
to see stock.
Add to basket
Description
Description
Compatible Microscopes:
  • Axio Examiner.D1
  • Axio Examiner.Z1
  • Axio Scope.A1
  • Axio Vert.A1 FL
  • Axio Vert.A1 MAT
Specifications
Specifications
Slider FL Attenuator
Filter wheel with 6 positions
14 x 40 mm
For insert into the aperture stop plane of reflected-light illuminator
1 Ea.