Läs mer
FLASH Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB500-219MFV450
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
FLASH Polyclonal antibody specifically detects FLASH in Human samples. It is validated for Western Blot
Specifikationer
| FLASH | |
| Polyclonal | |
| Western Blot | |
| CASP8 associated protein 2, caspase 8 associated protein 2, CED-4, FLASHKIAA1315CASP8-associated protein 2, FLICE associated huge, FLICE-associated huge protein, FLJ11208, human FLASH, RIP25FLASH homolog RIP25 | |
| C-terminus of the FLASH protein (proprietary peptide). | |
| 0.1 mL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 9994 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |