Learn More
Kartell™ Flask, Erlenmeyer, graduated polypropylene 24/29 500mL
Translusent in design to allow one to view the mixture with ease. Kartell™ Polypropylene Conical Erlenmeyer Flasks are a simple flask that has a variety of applications in both chemistry and biology.
Brand: Kartell™ 0146300
Additional Details : Weight : 0.10000kg
Description
Translusent in design to allow one to view the mixture with ease. Kartell™ Polypropylene Conical Erlenmeyer Flasks are a simple flask that has a variety of applications in both chemistry and biology.
- For general laboratory use
- Provided with standardized neck
- Autoclavable
- Suitable for foodstuff
Specifications
500 ml | |
500 mL | |
50 mL | |
Conical | |
Yes |
Translucent | |
Polypropylene | |
24/29 NS | |
Autoclavable |